Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Mouse IL4 (from E. coli)

Marketsource
Fournisseur:   Biorbyt
Mouse IL4 (from <i>E. coli</i>)
Fournisseur:  Biorbyt
Marketsource
Certificats

2 Options disponibles ci-dessous

Product Details & Documents

 

Spécifications de la famille de produits

Spécifications

Spécifications

  • Catalog No:
  • BIRBORB80062-5
  • BIRBORB80062-20
  • Nom de la protéine/du peptide:
  • IL4
  • IL4
  • Synonyme de protéine:
  • B cell growth factor 1
  • B cell growth factor 1
  • Species:
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • Sequence:
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S.
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S.
  • Activité biologique:
  • The ED50 as determined by the dose-dependant prolifiration of Murine HT-2C ells is 2.5 ng/ml.
  • The ED50 as determined by the dose-dependant prolifiration of Murine HT-2C ells is 2.5 ng/ml.
  • Purity:
  • > 95.0% as determined by RP-HPLC and SDS-PAGE
  • > 95.0% as determined by RP-HPLC and SDS-PAGE
  • Formulation:
  • Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Conditions de stockage:
  • Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Solubilité:
  • It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Tested applications:
  • SDS-PAGE , HPLC , FuncS
  • SDS-PAGE , HPLC , FuncS

Spécifications

Product Family Options

Informations sur le produit

  • Cdt
    Disponibilité
    Prix
  • 5 µG
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 5 µG
    1 * 5 µG

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    BIRBORB80062-5
    Nom de la protéine/du peptide
    IL4
    Synonyme de protéine
    B cell growth factor 1
    Species
    Mouse
    Source
    E. coli
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S.
    Activité biologique
    The ED50 as determined by the dose-dependant prolifiration of Murine HT-2C ells is 2.5 ng/ml.
    Purity
    > 95.0% as determined by RP-HPLC and SDS-PAGE
    Formulation
    Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
    Conditions de stockage
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
    Solubilité
    It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Tested applications
    SDS-PAGE , HPLC , FuncS
    Marketsource
  • 20 µG
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 20 µG
    1 * 20 µG

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    BIRBORB80062-20
    Nom de la protéine/du peptide
    IL4
    Synonyme de protéine
    B cell growth factor 1
    Species
    Mouse
    Source
    E. coli
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S.
    Activité biologique
    The ED50 as determined by the dose-dependant prolifiration of Murine HT-2C ells is 2.5 ng/ml.
    Purity
    > 95.0% as determined by RP-HPLC and SDS-PAGE
    Formulation
    Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
    Conditions de stockage
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
    Solubilité
    It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Tested applications
    SDS-PAGE , HPLC , FuncS
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.