Methylated DNA IP kit, BIOARRAY™
Methylated DNA IP kit, BIOARRAY™
BIOARRAY™ Methylated DNA IP kit, 10 Tests
Catalog # ENZOENZ450120010
Supplier:   Enzo Life Sciences
Methylated DNA IP kit, BIOARRAY™
Catalog # ENZOENZ450120010
Supplier:  Enzo Life Sciences
CAS Number:  
Methylated DNA IP kit, BIOARRAY™
BIOARRAY™ Methylated DNA IP kit, 10 Tests
CAS Number:  
Price
900,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
BIOARRAY™ Methylated DNA IP Kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis in the completion of diverse downstream applications.
  • Highly specific antibody is included to distinguish methylated DNA from pool
  • DNA enrichment for large-scale DNA methylation analysis
  • Easy to use with reproducible results
The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the capture and separation of methylated DNA from non-methylated DNA in only a few hours.
Delivery information: Kit contains Protein A Beads, Anti-5 methylcytosine, Control DNA, Control primer 1 , Control primer 2, IP Buffer, DNA denaturing Buffer, Control mouse DNA (methylated/nonmethylated), DNA elution buffer, magnetic rods.
Specifications
Specifications
  • Cdt:
    10 Tests
  • Désignation:
    BIOARRAY™ Methylated DNA IP kit