ContactUsLinkComponent
Sliding microtome, Slide 2003
Sliding microtome, Slide 2003
Catalog # FEAT030002
Supplier:  PFM Medical GmbH
Sliding microtome, Slide 2003
Catalog # FEAT030002
Supplier:  PFM Medical GmbH
Specifications
  • Désignation:
    Sliding microtome, Slide 2003
  • L×P×H:
    320×420×270 mm
  • Cdt:
    1
Specifications
About this item
The stable knife sliding action has unrivalled smoothness due to the horizontal cross-roller bearing. This results in less fatigue during daily use. The vertical object movement uses a cross-roller bearing system, is free from backlash and avoids vibration. The newly developed quick release system for both the universal cassette clamp and the standard object clamp in connection with the object orientation system and the user-friendly rotatable knife holder allows a quick and uncomplicated approach of the object to the blade. The section thickness can be set manually or automatically. The object coarse feed is controlled by the handwheel on the microtome.
  • An integrated finger guard provides the required protection in the area of the knife/blade
  • Easy to clean and maintain
  • Knife slide lock in three positions
  • Integrated protection tray
  • Section waste tray (optional)
  • Stable and distortion-free housing
  • Enclosed micrometer mechanism
  • Object orientation: X/y axes
  • Manual and automatic section thickness setting
The Slide 2003 is a powerful instrument for all paraffin work in routine practice, research and industry.
Delivery: Includes knife holder with integrated finger guard, removable protection tray at object holder and orientable universal cassette clamp.