Injector/detector plug nuts
Injector/detector plug nuts
Catalog # REST21883
Supplier: Restek
Jump to:
Product Details & Documents
These are used to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base.
- Used to cap off an injector to isolate leaks and a detector for thermal cleaning
- To check a detector or make-up gas flow rate
Specifications
- Désignation:Injector/detector plug nuts