GC Columns, Rxi® Guard/Retention Gap (Fused Silica)
Fournisseur: Restek
Certificats
À propos de cet article
Rxi® guard/retention gap columns are specially designed to be exceptionally inert.
- Extend column lifetime
- Excellent inertness - obtain lower detection limits for active compounds
- Sharper chromatographic peaks by utilising retention gap technology
- Maximum temperature: 360 °C
Frequently Bought Together