Solid phase extraction cartridges, Resprep®
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Cartouche, 6 ml, 1000 mg
N° de catalogue REST24034
Fournisseur:   Restek
Solid phase extraction cartridges, Resprep®
N° de catalogue REST24034
Fournisseur:  Restek
Solid phase extraction cartridges, Resprep®
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Cartouche, 6 ml, 1000 mg
Prix
274,00 €
Connexion Pour la disponibilité
1 * 30 ST
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Spécifications
  • Type de colonne:
    Cartouche
  • Capacité:
    6 ml
  • Désignation:
    SPE Florisil cartridges
  • Seq:
    03
  • Poids du lit:
    1000 mg