Order Entry
Belgium
ContactUsLinkComponent
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose, poudre blanche
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose, poudre blanche
Catalog # ICNA0215396801
Supplier:  MP Biomedicals
CAS Number:  
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose, poudre blanche
Catalog # ICNA0215396801
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Cdt:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Identity Test :
    Passes
  • Melting Point:
    132 - 142 °C
  • Optical Rotation (c=1, CHCl₃):
    +85° ± 5°

Specification Test Results

About this item

 :
 :462.45 g/mol
CAS Number:22224-41-5
Formula:C₂₆H₂₂O₈
MDL Number:MFCD00080818