ContactUsLinkComponent
Test kits, chlorine dioxide, Comparator 2000+
Test kits, chlorine dioxide, Comparator 2000+
Catalog # LOVI410001
Supplier:  Lovibond
CAS Number:  
undefined
Test kits, chlorine dioxide, Comparator 2000+
Catalog # LOVI410001
Supplier:  Lovibond
CAS Number:  
Specifications
  • Cdt:
    1 KIT
  • Désignation:
    Chlorine dioxide test kit
  • Type:
    AF 112ED
  • Plage de mesure:
    0,04 - 0,57 mg/l (ClO2)
Specifications
About this item
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Delivery: Test kit is supplied in a sturdy plastic case containing comparator 2000+, test discs, all the necessary cells, accessories and AQUALYTIC® tablet reagents (for 100 measurements).