Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Fournisseur: PHENOMENEX
Certificats
À propos de cet article
Luna® Phenyl-Hexyl provides separations not achievable on C18 or C8 columns; such as increased retention for polar, aromatic compounds as well as reversals in analyte elution order. Luna® Phenyl-Hexyl is a reproducible, extremely stable phenyl phase. Most phenyl phases use a short propyl (3 carbon) linker, which limits phase stability.
- Highly reproducible and stable phenyl phase
- Dual selectivity of both phenyl phase and a short alkyl phase (C5 or C8)
- Excellent retention of aromatic and polar, amine compounds
The Phenyl-Hexyl bonded phase employs a phenyl ring with a hexyl (6 carbon) linker and is densely bonded to Luna® silica surface. Dense bonding and the hexyl linker reduce bonded phase hydrolysis and increases chemical stability.
Spécifications
- Catalog No:
- PHEN00F-4257-E0
- PHEN00F-4256-E0
- PHEN00G-4257-E0
- PHEN00F-4256-B0
- Charge de carbone:
- 17,5%
- 17,5%
- 17,5%
- 17,5%
- End-capping:
- Oui
- Oui
- Oui
- Oui
- Plage de pH:
- 1,5 - 9,0
- 1,5 - 9,0
- 1,5 - 9,0
- 1,5 - 9,0
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- Code USP:
- L11
- L11
- L11
- L11