Nitrate de thallium 99.99% (sur base traces de métaux)
Catalog # ACRO194240100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder or crystals
- Assay:75.9 to 77.5 % (Tl)
- Total trace metal impurities:≤100 ppm
Specification Test Results
About this item
Synonyms: Thallium nitrate
:
:266.39 g/mol
Boiling Pt:433 °C (760 mmHg)
Melting Pt:206 °C
Density:1,273 g/cm³ (20 °C)
Storage Temperature:Température ambiante
UN:2727
ADR:6.1,II
:GHS06
:GHS08
:GHS09
:GHS03
CAS Number:10102-45-1
Formula:NO₃Tl
MDL Number:MFCD00011280