ContactUsLinkComponent
c-Myc Tag IP/Co-IP Kit, Pierce™
c-Myc Tag IP/Co-IP Kit, Pierce™
N° de catalogue PIER23620
Numéro CAS:  
c-Myc Tag IP/Co-IP Kit, Pierce™
N° de catalogue PIER23620
Numéro CAS:  
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    c-Myc-Tag IP/Co-IP kit, sufficient for 25 reactions
  • Seq:
    02
Spécifications
À propos de cet article
Pierce™ c-Myc Tag IP/Co-IP Kit provides a simple and fast method to study c-Myc-tagged proteins, with advantages over the traditional IP procedure using protein A/G agarose.
  • Immobilised anti-c-Myc monoclonal antibody binds the c-Myc epitope tag with high specificity, providing high yield immunoprecipitation products and clean Western blot detection
  • Excellent results with as little as 1 µg of anti-c-Myc antibody in IP mode with the positive control lysate
  • Kit includes a positive control lysate that contains over-expressed GST-c-Myc to validate the reagents and specific protocol, and the instructions provide tips for planning other controls
  • Affinity system is compatible with IP or Co-IP from various cell lysates and physiological (non-denaturing) buffer systems
  • Complete kit includes all necessary reagents, convenient spin columns, and easy-to-follow instructions
The high affinity anti-c-Myc antibody coupled agarose enables immunoprecipitation (IP) of c-Myc-tagged proteins or co-immunoprecipitation (Co-IP) of their interacting partners without antibody contamination. The Pierce™ spin columns provided with the kit handle small volumes of anti-c-Myc agarose and allow the entire procedure to be performed in a single spin column, preventing anti-cMyc agarose loss between washes. The spin columns further enhance recovery by efficiently separating buffer from the anti-c-Myc agarose.
Livraison: Kit contains sufficient components for 25 immunoprecipitations or co-immunoprecipitations involving c-Myc-tagged proteins, including 10 experiments with the c-Myc-tagged Positive Control.