Inclusion body solubilisation reagent, Pierce™
N° de catalogue PIER78115
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Spécifications
- Cdt:100 ml
- Désignation:Inclusion Body Solubilisation Reagent
Spécifications
À propos de cet article
Inclusion Body Solubilisation Reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents.
- Achieves the essential first step for subsequent insoluble protein refolding procedures
- One easy to use solution
- Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE may be performed
- Can accommodate additional components such as reducing agents or chelating agents