ContactUsLinkComponent
Pierce™, CarboxyLink™ Immobilisation Kit
  PIER44899
 :  
undefined
Pierce™, CarboxyLink™ Immobilisation Kit
  PIER44899
 :  
  • Cdt:
    1 KIT
  • Désignation:
    CarboxyLink™ immobilisation kit, 5×(2 ml, sufficient for immobilising 1 - 2 mg of peptide per column
CarboxyLink™ immobilisation kit is for covalent immobilisation of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.
  • Ideal for unmodified peptides, immobilises peptides with high capacity and various orientations without steric hindrance
  • DADPA-activated crosslinked 4% beaded agarose acting as a CarboxyLink™ coupling resin
  • With stable linkage, flexible and gentle coupling conditions
 : Kit includes CarboxyLink™ columns, 2 ml (5 columns), EDC crosslinker (5×60 mg), MES buffered saline pack (makes 500 ml), wash solution (120 ml), and column accessories.