Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Catalog # PIER34035
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Cdt:1 g
- Désignation:NBT (powder)
- Seq:01
Specifications
About this item
NBT (nitro-blue tetrazolium chloride) yields a blue-purple, water-insoluble product upon reduction of the tetrazolium salt. The product of reduction is NBT-formazan. This substrate is widely used for immunochemistry applications because the color is linear and stable. NBT is used as the substrate for both glucose oxidase and alkaline phosphatase systems.