Compat-Able™ Protein assay preparation reagent set
N° de catalogue PIER23215
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Spécifications
- Cdt:500 ml
- Désignation:Compat-Able™ Protein assay preparation reagent set, sufficient for treating up to 500 protein assay samples
Spécifications
À propos de cet article
The Compat-Able™ Protein assay preparation reagent set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
- Removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
- Prepare samples in less than 10 minutes with four simple steps
- Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
- Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
- Protocol integrates control and normalisation procedures
Livraison: Kit contains sufficient reagents to pretreat up to 500 samples. It comprises Compat-Able™ Protein assay preparation reagent 1 (250 ml) and Compat-Able™ Protein assay preparation reagent 2 (250 ml).