Carcass sampling kit
Carcass sampling kit
Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl), 200
N° de catalogue TSCLTS/15-BNACL
Fournisseur:   TSC LIMITED
Carcass sampling kit
N° de catalogue TSCLTS/15-BNACL
Fournisseur:  TSC LIMITED
Numéro CAS:  
Carcass sampling kit
Carcass sampling kit
Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl), 200
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 200 ST
Product Details & Documents
Carcass hygiene blue sampling sponge in easy open stomacher pouch dosed with 0.9% saline.
  • Blue cellulose sponge
  • Easy open stomacher pouch
  • Sterilised by gamma irradiation at dosage of 25-53.4 kGy
Recommended for:
Meat Carcass sampling when testing for Salmonella spp. Conforming to EC Commision 2073/2005 on foodstuffs & compliant with ISO 17604:2003
Certifications: EC 2073/2005 on foodstuffs and compliant with ISO 17604:2003.
Emballage: Individually packed & dosed with 0.9% Sodium Chloride (NaCl). 200 per box.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Spécifications
Spécifications
  • Cdt:
    200
  • Désignation:
    Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl)