Freund's incomplete adjuvant, Imject™
Freund's incomplete adjuvant, Imject™
Imject™ Freund's incomplete adjuvant, 5×10 ml, 1 SET
N° de catalogue PIER77145
Fournisseur:   Thermo Fisher Scientific
Freund's incomplete adjuvant, Imject™
N° de catalogue PIER77145
Numéro CAS:  
Freund's incomplete adjuvant, Imject™
Freund's incomplete adjuvant, Imject™
Imject™ Freund's incomplete adjuvant, 5×10 ml, 1 SET
Numéro CAS:  
Prix
145,00 €
Connexion Pour la disponibilité
1 * 1 SET
Product Details & Documents
Imject™ Freund's Incomplete Adjuvant (FIA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Water-in-oil emulsion without Mycobacterium
Livraison: Supplied ready to use in 10 ml ampoules.
Spécifications
Spécifications
  • Cdt:
    1 SET
  • Désignation:
    Imject™ Freund's incomplete adjuvant, 5×10 ml
Spécifications