Immunoprecipitation, magnetic ChIP kit, Pierce™
Catalog # PIER26157
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Magnetic ChIP Kit, sufficient for 30 ChIP assays
Specifications
About this item
The Pierce™ Magnetic ChIP Kit provides a convenient method for efficient isolation of chromatin-bound DNA by immunoprecipitation for subsequent quantitation by PCR.
- Purified DNA for quantitative PCR can be obtained in about 8 hours
- Optimised micrococcal nuclease digestion and nuclear lysis
- Obtain results with as few as 10000 cells
The blocked protein A/G magnetic beads provide high binding capacity, low non specific background, and work with many antibody species. These beads can be used manually with a magnetic stand or with automated platforms.
Delivery: The kit contains sufficient reagents to perform 30 chromatin immunoprecipitation (ChIP) assays with appropriate controls using an optimised protocol. The kit includes reagents for cell lysis, capture of protein-DNA complexes, reversal of crosslinking, and DNA isolation.