Net dressing for the head
CEDE1893
- Désignation:Grand pansement métallisé stérile et filet tubulaire sans latex pour maintenir le pansement en place
- Environmentally Preferable:
- Cdt:1
This kit contains a large sterile metallised dressing and latex-free tubular net intended for keeping the dressing in place. The dressing is intended to be used as a mechanical barrier and/or for absorption of exudates from large wounds on the head or other parts of the body, e.g. legs.
- Dressing: Absorbent layer made of non woven 100% viscose; non adherent layer of PE on both sides of dressing; aluminium metallised on one side
- Non adherent layer on both sides of the dressing prevents the dressing from sticking to the wound
- Metallised dressings are sterilised by irradiation
Dressing: 200×400 mm
Tubular net: 70×400 mm (unstretched)