ContactUsLinkComponent
Dimroth condensers with standard ground joints
Dimroth condensers with standard ground joints
N° de catalogue KAVA442517266
Fournisseur:  Kavalier
undefined
Dimroth condensers with standard ground joints
N° de catalogue KAVA442517266
Fournisseur:  Kavalier
Spécifications
  • Rodage mâle:
    29/32 NS
  • Longueur du condenseur:
    250 mm
  • Rodage femelle:
    29/32 NS
  • Cdt:
    1
Spécifications
À propos de cet article
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with core and jacket.