Conductivity standards
Conductivity standards
Conductivity standard, 12,88 mS/cm, 250 mL
N° de catalogue METT51350094
Fournisseur:   Mettler - Toledo
Conductivity standards
N° de catalogue METT51350094
Fournisseur:  Mettler - Toledo
Numéro CAS:  
Conductivity standards
Conductivity standards
Conductivity standard, 12,88 mS/cm, 250 mL
Numéro CAS:  
Prix
31,60 €
Connexion Pour la disponibilité
1 * 250 mL
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
Spécifications
Spécifications
  • Cdt:
    250 mL
  • Désignation:
    Conductivity standard
  • Conductivité:
    12,88 mS/cm
Spécifications