Microscope slides with printed rings
Microscopy slide with printed rings
N° de catalogue BELC5638-01920
Marketsource
Fournisseur: BELLCO GLASS
Microscope slides with printed rings
N° de catalogue BELC5638-01920
Fournisseur: BELLCO GLASS
Marketsource
Aller à :
Product Details & Documents
These microscopy slides with three printed rings allow for counting bacteria by spreading a known volume of solution over a known area for the calculation of bacteria per sample.
Area (ring): 1 cm²
Spécifications
- Désignation:Microscopy slide with printed rings