Anti-Ubiquitously expressed transcript Rabbit Polyclonal Antibody
100 μg
N° de catalogue BOSSBS-6749R
Fournisseur: Bioss
Anti-Ubiquitously expressed transcript Rabbit Polyclonal Antibody
N° de catalogue BOSSBS-6749R
Fournisseur: Bioss
Aller à :
Product Details & Documents
UXT is a novel protein, highly conserved in mouse. It interacts with the N terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is abundantly expressed in tumor tissues and is likely to be involved in tumorigenesis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Ubiquitously expressed transcript
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-6749R
Fiche technique
Spécifications
- Taille:100 μg
- Western blot:Yes
- Type d'anticorps:Primaire
- Formulaire:liquid
- Purification:Purified by Protein A.
- Synonymes antigène:ART 27|Protein UXT|Ubiquitously expressed transcript|UXT_HUMAN.|Androgen receptor trapped clone 27 protein|Ubiquitously expressed transcript protein
- Isotype:IgG
- Conjugaison:Unconjugated
- Concentration:1 ug/ul
- Réactivité:Human|Rat|Mouse
- ID de gène:8409
- Cat. No.:BOSSBS-6749R
- Clonalité:Polyclonal
- Immunogène:105-155/157
- Nom de l'antigène:Ubiquitously expressed transcript
- Hôte:Rabbit
- ImmunoChimie:Yes
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Température de transport:4 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



