Funnels, vacuum, according to Dr. Hirsch
Catalog # BIBB3766/03
Supplier: SciLabware
Specifications
- Pour:NS 14/23
- Ø ext. entonnoir sup.:35 mm
- Ø plaque:12 mm
- Porosité:3
- Cdt:1
Specifications
About this item
PYREX® borosilicate glass.
- Ideal for filtering semi-micro quantities of material
- Very good chemical resistance
- Very good temperature resistance
Hirsch pattern vacuum filtration funnel with sintered glass disc.