Solid phase extraction disks, Resprep® Oil & Grease
Solid phase extraction disks, Resprep® Oil & Grease, 47 mm
N° de catalogue REST26022
Fournisseur: Restek
Aller à :
Product Details & Documents
Glass fiber disks embedded with specialty bonded silica. These disks reduce the emulsion formation and amount of solvent required by EPA methods. No chlorofluorocarbons needed.
Certifications: Meets the requirements for EPA method 1664.
Spécifications
- Ø:47 mm
- Désignation:Solid phase extraction disks, Resprep® Oil & Grease