Wipes for industrial applications, Tork
SCAA128407
: Essity
- Pour:W1
- Emballage:1 roll of 340 m (1000 sheets)
- Couleur:bleu
- Désignation:Wipe roll, 3 ply, perforated and embossed
- L×l:340 m × 369 mm
- Cdt:1 Roll
These 3-ply wipes made of recycled fibres are ideal for industrial wiping tasks and hand wiping.
- Ideal for mopping
- Good oil and water absorption
- Suitable for hand wiping
- Food contact approved