Spray chamber
Spray chamber
N° de catalogue HEWLG1820-65337
Fournisseur: Agilent
Aller à :
Product Details & Documents
This spray chamber made of quartz is used with 7500 ICP-MS.
It is dedicated for babington, concentric and micro-concentric nebulisers.
Attention: Not suitable for hydrofluoric acid.
Spécifications
- Désignation:Spray chamber