Membrane disc filters, Optimizer®
Catalog # ENTECWAX04700
Supplier: ENTEGRIS
Specifications
- Ø:47 mm
- Taille de pore:0,02 µm
- Seq:01
- Cdt:1 SET
Specifications
About this item
These filter disc series can be used for simple, quick and efficient filtration of fluids used in laboratory, pilot and small scale applications.
- Used for clarification of acids, bases and solvents
- Used in cartridge filters to allow for consistent scale up of products