Electrochemistry kit
Equipment set for electrochemistry, 12 pieces
Catalog # BINH1002711
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
Equipment set inclusive of digital multimeter for measuring electrochemical potentials of various metals in experiments.
: Kit supplied with a digital multimeter with 2 cables with crocodile clamps, 1× flat trough, 1× copper plate, 1× zinc plate, 1× iron plate, 2× nickel plate, 1× aluminium plate, 2× electrolyte-carbon plates.
Specifications
- Désignation:Equipment set for electrochemistry, 12 pieces
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice