Order Entry
Belgium
ContactUsLinkComponent
Anti-RAB33A Mouse Polyclonal Antibody
Anti-RAB33A Mouse Polyclonal Antibody
N° de catalogue ABNOH00009363-B01P
Fournisseur:  Abnova
Anti-RAB33A Mouse Polyclonal Antibody
N° de catalogue ABNOH00009363-B01P
Fournisseur:  Abnova

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    RAB33A, member RAS oncogene family
  • Symbole de l'antigène:
    RAB33A
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Mouse
  • Western blot:
    Yes
  • Adsorption croisée:
    No
  • Format:
    Liquid
  • ID de gène:
    9363
  • Synonymes antigène:
    RabS10
  • Nombre d'acides aminés:
    1 to 237
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MAQPILGHGSLQPASAAGLASLELDSSLDQYVQIRIFKIIVIGDSNVGKTCLTFRFCGGTFPDKTEATIGVDFREKTVEIEGEKIKVQVWDTAGQERFRKSMVEHYYRNVHAVVFVYDVTKMTSFTNLKMWIQECNGHAVPPLVPKVLVGNKCDLREQIQVPSNLALKFADAHNMLLFETSAKDPKESQNVESIFMCLACRLKAQKSLLYRDAERQQGKVQKLEFPQEANSKTSCPC
  • Température de stockage:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogène:
    RAB33A (NP_004785.1, 1 a.a. ~ 237 a.a) full-length human protein.
  • Taille:
    50 μg
  • Cdt:
    50 µg

Spécifications

À propos de cet article

Mouse polyclonal antibody raised against a full-length human RAB33A protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: RAB33A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: