Anti-PSMD10 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12056
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Proteasome (prosome macropain) 26S Subunit non-ATPase 10
- Symbole de l'antigène:PSMD10
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Zebrafish,Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:No
- Format:Liquid
- ID de gène:5716
- Synonymes antigène:dJ889N15.2|p28(GANK)|p28
- Nombre d'acides aminés:100 to 200
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Température de stockage:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogène:A synthetic peptide corresponding to amino acids 100-200 of human PSMD10.
- Taille:100 μl
- Cdt:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of PSMD10.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: PSMD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat, Zebrafish