Anti-GPR120 Rabbit Polyclonal Antibody
Catalog # CAYM15130-1
Supplier: Cayman Chemical
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:GPR120
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Épitope:N-terminal
- Adsorption croisée:No
- Nombre d'acides aminés:28 to 40
- Storage buffer:TBS, pH 7,4, containing 5 mg/ml BSA
- Sequence:FSDVKGDHRLVLA
- Température de stockage:–20 °C
- Température de transport:–20 °C
- Immunogène:human GPR120 amino acids 28-40
- Purification:purified by affinity chromatography
- Taille:1 Ea
- Cdt:1
Specifications
About this item
Type: Primary
Antigen: GPR120
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human