Aspirating pipettes
Aspirating pipettes
2 ml
N° de catalogue GREI710183_1000
Fournisseur:   Greiner Bio-One
Aspirating pipettes
N° de catalogue GREI710183_1000
Fournisseur:  Greiner Bio-One
Aspirating pipettes
Aspirating pipettes
2 ml
Prix
201,00 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Free of detectable DNase, RNase, human DNA
  • Non pyrogenic, non cytotoxic
  • Maximum accuracy, drop-free pipetting
  • Maximum clarity from high-grade polystyrene
  • Lot number and best-before date on each bag
Emballage: Paper/plastic-packaging with peel-off function for individually wrapped pipettes with an additional break-through function.
Documents
Spécifications
  • Capacité:
    2 ml