Eye showers, free-standing
Eye showers, free-standing
Eye showers, free-standing
Fournisseur:  
Certificats
Eye showers, free-standing
Eye showers, free-standing
0 Options disponibles ci-dessous
Product Details & Documents
These free-standing eye showers feature two shower heads with a green epoxy painted stainless steel basin.
  • For installation on the ground
  • Made of stainless steel with grey epoxy paint
  • Available with hand pedal or with hand and foot pedal
Water connection: ½"
Water flow rate 17 L/min