Eye showers, free-standing
Aller à :
À propos de cet article
These free-standing eye showers feature two shower heads with a green epoxy painted stainless steel basin.
- For installation on the ground
- Made of stainless steel with grey epoxy paint
- Available with hand pedal or with hand and foot pedal
Water connection: ½"
Water flow rate 17 L/min
0 Options disponibles ci-dessous
Product Details & Documents
These free-standing eye showers feature two shower heads with a green epoxy painted stainless steel basin.
- For installation on the ground
- Made of stainless steel with grey epoxy paint
- Available with hand pedal or with hand and foot pedal
Water connection: ½"
Water flow rate 17 L/min
Product Family Options
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.