Chromogenic endotoxin testing reagents, Pyrochrome®
Catalog # CAPEC1500-5
Supplier: Associates of Cape Cod
CAS Number:
Specifications
- Cdt:300 Tests
- Désignation:Pyrochrome® Chromogenic test kit for 300 tests
Specifications
About this item
Pyrochrome® sets the standard for quality and redefines the way chromogenic reagents should perform. Pyrochrome is a versatile quantitative reagent that is used to perform kinetic or endpoint assays. It is a sensitive and flexible reagent that can be used for testing in compliance with the USP, EP and JP bacterial endotoxins test chapter. Pyrochrome is offered with either Pyrochrome Buffer or for endotoxin-specific testing, with Glucashield® Buffer. Pyrochrome can be used with the new Pyros Kinetix® Flex tube reader at a 1:1 and economical 4:1 ratio.
- Can achieve a maximum sensitivity of 0,001 EU/ml
- Highly cost effective chromogenic reagent