- Type d'anticorps:Secondaire
- Nom de l'antigène:Immunoglobulin G
- Symbole de l'antigène:IgG
- Clonalité:Polyclonal
- Conjugaison:DyLight® 594
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Goat
- Western blot:Yes
- Adsorption croisée:No
- Format:Purified IgG
- Formulaire:Lyophilized
- Storage buffer:10 mM Sodium phosphate, 0,15 M Sodium chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
- Température de stockage:2 - 8 °C
- Concentration:1,0 mg/ml (E 1% at 280 nm = 13,0)
- Température de transport:2 - 8 °C
- Immunogène:Goat IgG
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phase Goat IgG
- Taille:1,0 mg
- Cdt:1 mg
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593nm / 618nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat