Specifications
- Type d'anticorps:Secondaire
- Nom de l'antigène:Immunoglobulin G
- Symbole de l'antigène:IgG
- Clonalité:Polyclonal
- Conjugaison:AP (Alkaline Phosphatase)
- ELISA:Yes
- Cytométrie en flux:Yes
- Changement de gel:Yes
- Hôte:Goat
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Mouse
- Western blot:Yes
- Adsorption croisée:Yes
- Format:Purified IgG
- Formulaire:Clear, colorless liquid
- Fragment d'anticorps:F(ab')2
- Storage buffer:30 mM Triethanolamine, pH 7,2, 5 mM Magnesium Chloride, 0,1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
- Température de stockage:2…8 °C
- Concentration:1,50 mg/ml (E 1% at 280 nm = 13,0)
- Température de transport:2…8 °C
- Immunogène:Mouse IgG
- Purification:Affinity purified using solid phaseMouse IgG
- Taille:0,5 mg
- Cdt:0,5 mg
Specifications
About this item
ImmunoReagents Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
- Origin: US
ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse