- Type d'anticorps:Secondaire
- Nom de l'antigène:Immunoglobulin M mu-chain
- Symbole de l'antigène:IgM
- Clonalité:Polyclonal
- Conjugaison:Biotin (NHS-LC)
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Goat
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgM
- Réactivité:Human
- Western blot:Yes
- Épitope:µ-chain
- Adsorption croisée:No
- Format:Purified IgG
- Formulaire:Clear, colorless liquid
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
- Température de stockage:2…8 °C
- Concentration:1,50 mg/ml (E 1% at 280 nm = 13,0)
- Température de transport:2…8 °C
- Immunogène:Human IgM, µ chain
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phase Human IgM
- Taille:1,0 mg
- Cdt:1 mg
ImmunoReagents Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.
- Origin: US
ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Human