Order Entry
Belgium
ContactUsLinkComponent
Anti-IgA alpha Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgA alpha Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
N° de catalogue IMMRIR1509
Fournisseur:  ImmunoReagents
undefined
Anti-IgA alpha Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
N° de catalogue IMMRIR1509
Fournisseur:  ImmunoReagents

Spécifications

  • Type d'anticorps:
    Secondaire
  • Nom de l'antigène:
    Immunoglobulin A Alpha
  • Symbole de l'antigène:
    IgA
  • Clonalité:
    Polyclonal
  • Conjugaison:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Changement de gel:
    Yes
  • Hôte:
    Goat
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Précipitation immunologique:
    Yes
  • Isotype:
    IgA
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Adsorption croisée:
    No
  • Format:
    Purified IgG
  • Formulaire:
    Clear, colorless liquid
  • Storage buffer:
    30 mM Triethanolamine, pH 7,2, 5 mM Magnesium Chloride, 0,1 mM Zinc Chloride, 1% (w/v) BSA, Protease/IgG free
  • Température de stockage:
    2...8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Température de transport:
    2...8 °C
  • Immunogène:
    Human IgA, (α chain)
  • Purification:
    Affinity purified using solid phase Human IgA
  • Taille:
    1,0 mg
  • Cdt:
    1 mg

Spécifications

À propos de cet article

ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes Colour in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Informations de commande: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human