Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
IMMRIR1280
- Type d'anticorps:Secondaire
- Nom de l'antigène:Immunoglobulin G
- Symbole de l'antigène:IgG
- Clonalité:Polyclonal
- Conjugaison:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Cytométrie en flux:Yes
- Changement de gel:Yes
- Hôte:Donkey
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Rat
- Western blot:Yes
- Adsorption croisée:Yes
- Format:Purified IgG
- Formulaire:Lyophilised
- Fragment d'anticorps:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
- Température de stockage:2...8 °C
- Concentration:1,0 mg/ml (E 1% at 280 nm = 13,0)
- Température de transport:2...8 °C
- Immunogène:Rat IgG
- Purification:Affinity purified using solid phaseRat IgG
- Taille:0,5 mg
- Cdt:0,5 mg
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
: Products can by customized based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rat