Anti-CD70 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H173]
Catalog # USBIC2425-01N
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CD70 molecule
- Symbole de l'antigène:CD70
- Clonalité:Monoclonal
- Clone:7H173
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- Isotype:IgG1
- Réactivité:Human
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2, 5% glycerol, 0,2% BSA, 0,05% sodium azide.
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Taille:120 Tests
- Cdt:120 Tests
Specifications
About this item
Type: Primary
Antigen: CD70 (CD70 molecule)
Clonality: Monoclonal
Clone: 7H173
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human