Anti-ZNF557 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI044280-FITC
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:zinc finger protein 557
- Symbole de l'antigène:ZNF557
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:N-terminal
- Formulaire:liquid
- Nombre d'acides aminés:91 to 118
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ZNF557 (zinc finger protein 557)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human