Anti-KLDC2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI037518-PE
- Type d'anticorps:Primaire
- Symbole de l'antigène:KLDC2
- Clonalité:Polyclonal
- Conjugaison:PE (Phycoerythrin)
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Épitope:C-terminal
- Formulaire:liquid
- Nombre d'acides aminés:299 to 329
- Storage buffer:PBS, pH 7,2
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: KLDC2
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse