Anti-His tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # GENSA00612
Supplier: Genscript
Specifications
- Type d'anticorps:Tag
- Symbole de l'antigène:His
- Clonalité:Monoclonal
- Conjugaison:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Hôte:Mouse
- Isotype:IgG1
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Type: Tag
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: