Anti-SLC2A9 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-06271F
Supplier: Novus Biologicals
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:solute carrier family 2 (facilitated glucose transporter), member 9
- Symbole de l'antigène:Solute carrier family 2, member 9
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- Réactivité:Mouse
- Western blot:Yes
- ID de gène:56606
- Synonymes antigène:GLUTX|UAQTL2|GLUT9|URATv1
- Température de stockage:Cool
- Température de transport:Ice
- Taille:100 μL
- Cdt:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC2A9 (solute carrier family 2 (facilitated glucose transporter), member 9)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse