About this item
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : COPS5 (sous-unité 5 du signalosome COP9)
Clonalité : Polyclonal
Clone:
Conjugaison : Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité : Humain
Specifications
- Catalog No:
- ABCAAB74271-200
- Symbole de l'antigène:
- COPS5
- Nom de l'antigène:
- COP9 signalosome subunit 5
- Type d'anticorps:
- Primaire
- Clonalité:
- Polyclonal
- Conjugaison:
- Unconjugated
- Hôte:
- Rabbit
- Western blot:
- Yes
- ImmunoChimie:
- Yes
- Température de stockage:
- Cool
- Température de transport:
- Ice