Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Bacillus anthracis lethal factor
- Symbole de l'antigène:B. anthracis LF
- Clonalité:Monoclonal
- Clone:LF2
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Mouse
- Précipitation immunologique:Yes
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:100 µG
Spécifications
À propos de cet article
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : B. anthracis LF (facteur létal de Bacillus anthracis)
Clonalité : Monoclonal
Clone : LF2
Conjugaison: Non conjugué
Epitope :
Hôte : Souris
Isotype: IgG1
Réactivité: