Order Entry
Belgium
ContactUsLinkComponent
Anti-Matriptase 2 Rabbit Polyclonal Antibody
Anti-Matriptase 2 Rabbit Polyclonal Antibody
  ABCAAB56180-100
 :  Abcam
Anti-Matriptase 2 Rabbit Polyclonal Antibody
  ABCAAB56180-100
 :  Abcam

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Matriptase 2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Précipitation immunologique:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    cytoplasmic domain
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Matriptase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: IgG
Reactivity: Human