Anti-Poliovirus type 3 Mouse Monoclonal Antibody [clone: 300-06]
N° de catalogue ABCAAB47803-100
Fournisseur: Abcam
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Poliovirus Type 3
- Symbole de l'antigène:PV3
- Clonalité:Monoclonal
- Clone:300-06
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Mouse
- Isotype:IgG2a
- Neutralisation:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
Attention:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Poliovirus type 3
Clonality: monoclonal
Clone: 300-06
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: