Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Tumor protein p53
- Symbole de l'antigène:P53
- Clonalité:Monoclonal
- Clone:E26
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:100 µl
Specifications
About this item
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : TP53 (protéine tumorale p53)
Clonalité : Monoclonal
Clone : E26
Conjugaison: Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité :